Anti-CD45R Rat monoclonal antibody APC (Allophycocyanin)/Cy7® [clone: RA3-6B2]
Proveedor: Abcam
ABCAAB79107-50EA 530 EUR
ABCAAB79107-50
Anti-CD45R Rat monoclonal antibody APC (Allophycocyanin)/Cy7® [clone: RA3-6B2]
Type: Primary
Antigen: CD45R (CD45R)
Clonality: monoclonal
Clone: RA3-6B2
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse