Anti-LYPLAL1 Goat Polyclonal Antibody
Proveedor: Abcam
ABCAAB77640-200EA 510 EUR
ABCAAB77640-200
Anti-LYPLAL1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LYPLAL1 (lysophospholipase-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse