Anti- IgG Antibody (Biotin)

Proveedor: Abcam
ABCAAB64257-125EA 635 EUR
ABCAAB64257-125
Anti- IgG Antibody (Biotin)
Type: Secondary
Antigen: Mouse + Rabbit IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR