Anti-TIMP1 Rabbit polyclonal antibody

Proveedor: Abcam
ABCAAB61224-100EA 560 EUR
ABCAAB61224-100
Anti-TIMP1 Rabbit polyclonal antibody
Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR