Anti-TOMM22 Mouse Monoclonal Antibody

Proveedor: Abcam
ABCAAB57523-100EA 575 EUR
ABCAAB57523-100
Anti-TOMM22 Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: TOMM22 (translocase of outer mitochondrial membrane 22 homolog (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR