Anti-AMELX Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB54507-100EA 530 EUR
ABCAAB54507-100
Anti-AMELX Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: AMELX (amelogenin, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now