Anti-MCM7 Rabbit Monoclonal Antibody [clone: EP1974Y]

Proveedor: Abcam
ABCAAB52489-100EA 832 EUR
ABCAAB52489-100
Anti-MCM7 Rabbit Monoclonal Antibody [clone: EP1974Y]
Anticuerpos
Type: Primary
Antigen: MCM7 (minichromosome maintenance complex component 7)
Clonality: monoclonal
Clone: EP1974Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR