Anti-NOVA2 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB51004-100EA 739 EUR
ABCAAB51004-100
Anti-NOVA2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NOVA2 (neuro-oncological ventral antigen 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human