Anti-ASH1L Mouse monoclonal antibody unconjugated [clone: ASH5H03]
Proveedor: Abcam
ABCAAB50981-100EA 495 EUR
ABCAAB50981-100
Anti-ASH1L Mouse monoclonal antibody unconjugated [clone: ASH5H03]
Anticuerpos
Type: Primary
Antigen: ASH1L (ash1 (absent, small, or homeotic)-like (Drosophila))
Clonality: monoclonal
Clone: ASH5H03
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now