Anti-CSRNP1 Mouse monoclonal antibody unconjugated [clone: 2055E3a]
Proveedor: Abcam
ABCAAB50978-100EA 510 EUR
ABCAAB50978-100
Anti-CSRNP1 Mouse monoclonal antibody unconjugated [clone: 2055E3a]
Anticuerpos
Type: Primary
Antigen: CSRNP1 (cysteine-serine-rich nuclear protein 1)
Clonality: monoclonal
Clone: 2055E3a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human