Anti-YARS Mouse Monoclonal Antibody [clone: YAR5H08]
Proveedor: Abcam
ABCAAB50961-100EA 550 EUR
ABCAAB50961-100
Anti-YARS Mouse Monoclonal Antibody [clone: YAR5H08]
Anticuerpos
Type: Primary
Antigen: YARS (tyrosyl-tRNA synthetase)
Clonality: monoclonal
Clone: YAR5H08
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat