Anti-RPA1 Mouse monoclonal antibody unconjugated [clone: RPA15I373]
ABCAAB50936-100EA 720 EUR
ABCAAB50936-100
Anti-RPA1 Mouse monoclonal antibody unconjugated [clone: RPA15I373]
Type: Primary
Antigen: RPA1 (replication protein A1, 70kDa)
Clonality: monoclonal
Clone: RPA15I373
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human