Anti-SFPQ Rabbit polyclonal antibody unconjugated
ABCAAB50935-100EA 737 EUR
ABCAAB50935-100
Anti-SFPQ Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: SFPQ (splicing factor proline/glutamine-rich)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human