Anti-GATA1 Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB28839-100EA 575 EUR
ABCAAB28839-100
Anti-GATA1 Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: GATA1 (GATA binding protein 1 (globin transcription factor 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR