Anti-Scavenging Receptor SR-BI Rabbit Polyclonal Antibody

ABCAAB24603-100EA 716 EUR
ABCAAB24603-100
Anti-Scavenging Receptor SR-BI Rabbit Polyclonal Antibody
Type: Primary
Antigen: Scavenging Receptor SR-BI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR