Anti-MC4R Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB24233-100EA 815 EUR
ABCAAB24233-100
Anti-MC4R Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MC4R (melanocortin 4 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR