Anti-Blood Group H ab antigen Mouse Monoclonal Antibody [clone: 87-N]

Proveedor: Abcam
ABCAAB24222-100EA 781 EUR
ABCAAB24222-100
Anti-Blood Group H ab antigen Mouse Monoclonal Antibody [clone: 87-N]
Anticuerpos
Type: Primary
Antigen: Blood Group H ab antigen
Clonality: monoclonal
Clone: 87-N
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR