Anti-HIV1 Goat polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB24209-250EA 703 EUR
ABCAAB24209-250
Anti-HIV1 Goat polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: HIV1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR