Anti-COL1/3 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB24135-100EA 781 EUR
ABCAAB24135-100
Anti-COL1/3 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: COL1/3 (collagen, type I/III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR