Anti-ABCC5 Rat Monoclonal Antibody [clone: M5II-54]

Proveedor: Abcam
ABCAAB24107-500EA 530 EUR
ABCAAB24107-500
Anti-ABCC5 Rat Monoclonal Antibody [clone: M5II-54]
Anticuerpos
Type: Primary
Antigen: ABCC5 (ATP-binding cassette, sub-family C (CFTR/MRP), member 5)
Clonality: monoclonal
Clone: M5II-54
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR