Anti-Sodium / Hydrogen Exchanger 1 Mouse Monoclonal Antibody [clone: 4E9]
Proveedor: Abcam
ABCAAB24018-50EA 525 EUR
ABCAAB24018-50
Anti-Sodium / Hydrogen Exchanger 1 Mouse Monoclonal Antibody [clone: 4E9]
Anticuerpos
Type: Primary
Antigen: Sodium / Hydrogen Exchanger 1
Clonality: monoclonal
Clone: 4000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig