Anti-RASSF1a Mouse Monoclonal Antibody [clone: 3F3]
Proveedor: Abcam
ABCAAB23950-100EA 495 EUR
ABCAAB23950-100
Anti-RASSF1a Mouse Monoclonal Antibody [clone: 3F3]
Anticuerpos
Type: Primary
Antigen: RASSF1a
Clonality: monoclonal
Clone: 3F3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse