Anti-TIMM50 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB23938-100EA 560 EUR
ABCAAB23938-100
Anti-TIMM50 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TIMM50 (translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR