Anti-PTPRC Mouse monoclonal antibody [clone: K252.1E4]

ABCAAB23918-1EA 588 EUR
ABCAAB23918-1
Anti-PTPRC Mouse monoclonal antibody [clone: K252.1E4]
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: K252.1E4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR