Anti-Epoxide hydrolase Goat polyclonal antibody

ABCAAB23850-100EA 604 EUR
ABCAAB23850-100
Anti-Epoxide hydrolase Goat polyclonal antibody
Type: Primary
Antigen: Epoxide hydrolase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR