Anti-HLA B27 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HLA-ABC-m3]

Proveedor: Abcam
ABCAAB23840-250EA 0 EUR
ABCAAB23840-250
Anti-HLA B27 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HLA-ABC-m3]
Anticuerpos
Type: Primary
Antigen: HLA B27
Clonality: monoclonal
Clone: HLA-ABC-m3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR