Anti-DAD1 Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB23836-50EA 799 EUR
ABCAAB23836-50
Anti-DAD1 Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: DAD1 (defender against cell death 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR