Anti-PTGES Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB23727-100EA 770 EUR
ABCAAB23727-100
Anti-PTGES Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PTGES (prostaglandin E synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR