Anti-Phospholipase A2 Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB23705-100EA 805 EUR
ABCAAB23705-100
Anti-Phospholipase A2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Phospholipase A2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR