Anti-LTA4H Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB23677-100EA 786 EUR
ABCAAB23677-100
Anti-LTA4H Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LTA4H (leukotriene A4 hydrolase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR