Anti-ACADM Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB23675-300EA 739 EUR
ABCAAB23675-300
Anti-ACADM Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ACADM (acyl-CoA dehydrogenase, C-4 to C-12 straight chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR