Anti-PPARG Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB23673-100EA 555 EUR
ABCAAB23673-100
Anti-PPARG Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PPARG (peroxisome proliferator-activated receptor gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR