Anti-HNRNPK Mouse Monoclonal Antibody [clone: F45 P9 C7]

Proveedor: Abcam
ABCAAB23644-100EA 808 EUR
ABCAAB23644-100
Anti-HNRNPK Mouse Monoclonal Antibody [clone: F45 P9 C7]
Anticuerpos
Type: Primary
Antigen: HNRNPK (heterogeneous nuclear ribonucleoprotein K)
Clonality: monoclonal
Clone: F45 P9 C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR