Anti-CD28 Mouse Monoclonal Antibody [clone: 204.12]

Proveedor: Abcam
ABCAAB23627-100EA 739 EUR
ABCAAB23627-100
Anti-CD28 Mouse Monoclonal Antibody [clone: 204.12]
Anticuerpos
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone: 204.12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR