Anti-Nectin 2 Mouse Monoclonal Antibody [clone: R2.525]

Proveedor: Abcam
ABCAAB23601-100EA 739 EUR
ABCAAB23601-100
Anti-Nectin 2 Mouse Monoclonal Antibody [clone: R2.525]
Anticuerpos
Type: Primary
Antigen: Nectin 2
Clonality: monoclonal
Clone: R2.525
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR