Anti-CD200 / OX2 Mouse Monoclonal Antibody [clone: OX-104]

Proveedor: Abcam
ABCAAB23552-100EA 773 EUR
ABCAAB23552-100
Anti-CD200 / OX2 Mouse Monoclonal Antibody [clone: OX-104]
Anticuerpos
Type: Primary
Antigen: CD200 / OX2
Clonality: monoclonal
Clone: OX-104
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR