Anti-PXN Mouse Monoclonal Antibody [clone: M107]

Proveedor: Abcam
ABCAAB23510-100EA 550 EUR
ABCAAB23510-100
Anti-PXN Mouse Monoclonal Antibody [clone: M107]
Anticuerpos
Type: Primary
Antigen: PXN (paxillin)
Clonality: monoclonal
Clone: M107
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR