Anti-C4A Mouse monoclonal antibody [clone: 25.3F4]

ABCAAB23473-100EA 611 EUR
ABCAAB23473-100
Anti-C4A Mouse monoclonal antibody [clone: 25.3F4]
Type: Primary
Antigen: C4A (complement component 4A (Rodgers blood group))
Clonality: monoclonal
Clone: 25.3F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR