Anti-Mannan Binding Lectin Mouse Monoclonal Antibody [clone: 3B6]

Proveedor: Abcam
ABCAAB23457-100EA 0 EUR
ABCAAB23457-100
Anti-Mannan Binding Lectin Mouse Monoclonal Antibody [clone: 3B6]
Anticuerpos
Type: Primary
Antigen: Mannan Binding Lectin
Clonality: monoclonal
Clone: 3B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR