Anti-VTN Mouse Monoclonal Antibody [clone: A18]

Proveedor: Abcam
ABCAAB23444-100EA 781 EUR
ABCAAB23444-100
Anti-VTN Mouse Monoclonal Antibody [clone: A18]
Anticuerpos
Type: Primary
Antigen: VTN (vitronectin)
Clonality: monoclonal
Clone: A18
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR