Anti-Nocturnin Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB22850-100EA 703 EUR
ABCAAB22850-100
Anti-Nocturnin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Nocturnin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR