Anti-BAZ2B Goat polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB22821-100EA 739 EUR
ABCAAB22821-100
Anti-BAZ2B Goat polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: BAZ2B (bromodomain adjacent to zinc finger domain, 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR