Anti-PUF60 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB22819-100EA 805 EUR
ABCAAB22819-100
Anti-PUF60 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PUF60 (poly-U binding splicing factor 60KDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR