Anti-MDM2 Mouse Monoclonal Antibody [clone: 2G2]

Proveedor: Abcam
ABCAAB22710-50EA 0 EUR
ABCAAB22710-50
Anti-MDM2 Mouse Monoclonal Antibody [clone: 2G2]
Anticuerpos
Type: Primary
Antigen: MDM2 (MDM2 oncogene, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: 2G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR