Anti-Proteasome 20S alpha 3 Mouse monoclonal antibody [clone: MCP72]

ABCAAB22667-50EA 588 EUR
ABCAAB22667-50
Anti-Proteasome 20S alpha 3 Mouse monoclonal antibody [clone: MCP72]
Type: Primary
Antigen: Proteasome 20S alpha 3
Clonality: monoclonal
Clone: MCP72
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR