Anti-PYY Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB22663-50EA 838 EUR
ABCAAB22663-50
Anti-PYY Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PYY (peptide YY)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR