Anti-Proteasome subunit beta type 2 Mouse Monoclonal Antibody [clone: MCP165]

Proveedor: Abcam
ABCAAB22650-50EA 575 EUR
ABCAAB22650-50
Anti-Proteasome subunit beta type 2 Mouse Monoclonal Antibody [clone: MCP165]
Anticuerpos
Type: Primary
Antigen: Proteasome subunit beta type 2
Clonality: monoclonal
Clone: MCP165
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR