Anti-Met Enkephalin Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB22620-50EA 788 EUR
ABCAAB22620-50
Anti-Met Enkephalin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Met Enkephalin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR