Anti-Interferon gamma Mouse monoclonal antibody [clone: DB-1]

ABCAAB22543-125EA 588 EUR
ABCAAB22543-125
Anti-Interferon gamma Mouse monoclonal antibody [clone: DB-1]
Type: Primary
Antigen: Interferon gamma
Clonality: monoclonal
Clone: DB-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR