Anti-ORC6 Rat Monoclonal Antibody [clone: 3A4]

Proveedor: Abcam
ABCAAB22536-50EA 703 EUR
ABCAAB22536-50
Anti-ORC6 Rat Monoclonal Antibody [clone: 3A4]
Anticuerpos
Type: Primary
Antigen: ORC6 (origin recognition complex, subunit 6)
Clonality: monoclonal
Clone: 3A4
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR